missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitrotoluene, 99%

Product Code. 11498877
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498877 10 g 10g
11404070 5 g 5g
11408887 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498877 Supplier Thermo Scientific Alfa Aesar Supplier No. B20520.09
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29682-46-0 | C7H5Cl2NO2 | 206.02 g/mol

2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29682-46-0
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00024185
InChI Key WBNZUUIFTPNYRN-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
PubChem CID 97155
IUPAC Name 1,3-dichloro-2-methyl-4-nitrobenzene
SMILES CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O

Specifications

Melting Point 53°C to 55°C
Quantity 10 g
UN Number UN2811
Beilstein 2259855
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 99%
Chemical Name or Material 2,6-Dichloro-3-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.