missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-fluorophenol, 98%

Product Code. 11493267
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493267 100 g 100g
11473267 5 g 5g
11483267 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493267 Supplier Thermo Scientific Alfa Aesar Supplier No. A16193.22
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 392-71-2 | C6H3Cl2FO | 180.987 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-71-2
Molecular Formula C6H3Cl2FO
Molecular Weight (g/mol) 180.987
MDL Number MFCD00010675
InChI Key BOJVIFKSTRCIRJ-UHFFFAOYSA-N
PubChem CID 94950
IUPAC Name 2,6-dichloro-4-fluorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)F

Specifications

Melting Point 47°C to 51°C
Boiling Point 142°C (100 mmHg)
Flash Point 103°C (217°F)
Odor Phenol-like
Quantity 100 g
UN Number UN3261
Beilstein 2046856
Synonym phenol, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluoro-phenol, pubchem2871, acmc-1ai43, 4-fluoro-2,6-dichlorophenol, bojvifkstrcirj-uhfffaoysa, phenol,2,6-dichloro-4-fluoro, attercop-chm at126185
Formula Weight 180.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-4-fluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.