missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethoxy)aniline, 96%, Thermo Scientific™

Product Code. 11995221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11995221 5 g 5g
11985221 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995221 Supplier Thermo Scientific Acros Supplier No. 442300050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 99479-66-0
Molecular Formula C7H4Cl2F3NO
Molecular Weight (g/mol) 246.01
MDL Number MFCD00190128
InChI Key FKISQWQHZULEEG-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethoxy aniline, benzenamine, 2,6-dichloro-4-trifluoromethoxy, 2,6-dichloro-4-trifluoromethoxy phenylamine, pubchem8461, acmc-209scs, 3.5-dichloro-4-amino trifluoromethoxybenzene, 2,6-dichloro-4-trifluoromethoxy-phenylamine, 3,5-dichloro-4-amino trifluoromethoxybenzene, 3,5-dichloro-4-aminotrifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy benzenamine
PubChem CID 688247
IUPAC Name 2,6-dichloro-4-(trifluoromethoxy)aniline
SMILES NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl

Tekniske data

CAS Min % 95.0
CAS Max % 100.0
Melting Point 32°C to 34°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Quantity 5 g
Formula Weight 246.02
Percent Purity 96%
Physical Form Powder, Low Melting Solid or Crystalline Needles
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethoxy)aniline
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 2,6-Dichloro-4-(trifluoromethoxy)aniline
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.