missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 11475586
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475586 5 g 5g
11465586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475586 Supplier Thermo Scientific Alfa Aesar Supplier No. B20728.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24279-39-8
Molecular Formula C7H4Cl2F3N
Molecular Weight (g/mol) 230.01
MDL Number MFCD00052918
InChI Key ITNMAZSPBLRJLU-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl aniline, 4-amino-3,5-dichlorobenzotrifluoride, 3,5-dichloro-4-amino benzotrifluoride, 3,5-dichloro-4-aminobenzotrifluoride, 2,6-dichloro-4-trifluoromethyl benzenamine, benzenamine, 2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl phenylamine, 2,6-dichloro-4-trifluoromethyl-benzenamine, pubchem2187
PubChem CID 141094
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F

Specifications

Melting Point 34°C to 37°C
Density 1.532
Boiling Point 60°C to 62°C (1 mmHg)
Flash Point 88°C (190°F)
Quantity 5 g
UN Number UN3077
Beilstein 2838819
Formula Weight 230.02
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)aniline
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.