missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10497242
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10497242 1 g Amber Glass Bottle 1g
10293512 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10497242 Supplier Thermo Scientific Maybridge Supplier No. SEW01637DA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 157021-61-9
Molecular Formula C8H2Cl2F3N
Molecular Weight (g/mol) 240.006
MDL Number MFCD00220879
InChI Key NCXSSFQXQAOREM-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl benzonitrile, 4-cyano-3,5-dichlorobenzotrifluoride, benzonitrile,2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl benzenecarbonitrile, buttpark 14550-33, intermediates-zcf02189, ncxssfqxqaorem-uhfffaoysa, buttpark 145\50-33
PubChem CID 2736870
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)benzonitrile
SMILES C1=C(C=C(C(=C1Cl)C#N)Cl)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 240.01
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.