missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoyl chloride, 98%

Product Code. p-104375 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10216050

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4659-45-4 | C7H3Cl3O | 209.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4659-45-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.46
MDL Number MFCD00000662
InChI Key JBLIDPPHFGWTKU-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
PubChem CID 78392
IUPAC Name 2,6-dichlorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 18°C
Color Colorless to Yellow or Pink
Density 1.4500g/mL
Boiling Point 142°C to 143°C (21.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3COCl
Refractive Index 1.5590 to 1.561
Quantity 10 mL
Beilstein 09, 343
Specific Gravity 1.45
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone,slightly soluble in heptane
Formula Weight 209.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichlorobenzoyl chloride, 99%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.