missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetonitrile, 97%, Thermo Scientific™

Product Code. 10133110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
25g; Glass bottle
Product Code.
10133110 25g; Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10133110

Brand: Thermo Scientific Acros 155080250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3215-64-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 74.0°C to 77.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H5Cl2N
Linear Formula Cl2C6H3CH2CN
MDL Number MFCD00001901
Quantity 25 g
Synonym 2,6-dichlorophenylacetonitrile, 2,6-dichlorobenzyl cyanide, 2-2,6-dichlorophenyl acetonitrile, 2,6-dichlorophenyl acetonitrile, 2,6-dichlorobenzeneacetonitrile, benzeneacetonitrile, 2,6-dichloro, acetonitrile, 2,6-dichlorophenyl, 2,6-dichloro-phenyl-acetonitrile, 2-2,6-dichlorophenyl ethanenitrile
InChI Key AOEJUUCUKRUCEF-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)CC#N)Cl
IUPAC Name 2-(2,6-dichlorophenyl)acetonitrile
Molecular Weight (g/mol) 186.04
PubChem CID 76678
Formula Weight 186.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dichlorophenylacetonitrile, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.