missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylthiourea, TRC

Product Code. 30384819
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384819 5 g 5g
30380483 10 g -
30370553 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384819 Supplier Toronto Research Chemicals Supplier No. TRCD6504605G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6590-91-6
Molecular Formula C7 H6 Cl2 N2 S
Molecular Weight (g/mol) 221.11
Synonym 1-(2,6-Dichlorophenyl)thiourea
IUPAC Name (2,6-dichlorophenyl)thiourea
SMILES NC(=S)Nc1c(Cl)cccc1Cl

Specifikationer

Quantity 5 g
InChI Formula InChI=1S/C7H6Cl2N2S/c8-4-2-1-3-5(9)6(4)11-7(10)12/h1-3H,(H3,10,11,12)
Formula Weight 219.9629
Chemical Name or Material 1-(2,6-Dichlorophenyl)thiourea
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.