missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorostyrene, 96%, stab. with 0.1% 4-tert-butylcatechol
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.
Solubility
Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.
Solubility
Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 1.266 |
| Boiling Point | 88°C to 90°C (8 mmHg) |
| Flash Point | 71°C (159°F) |
| Refractive Index | 1.574 |
| Quantity | 5 g |
| Beilstein | 1931527 |
| Solubility Information | Immiscible with water. |
| Formula Weight | 173.04 |
| Percent Purity | 96% |
| Chemical Name or Material | 2,6-Dichlorostyrene, stabilized with 0.1% 4-tert-butylcatechol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction