missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorostyrene, 96%, stab. with 0.1% 4-tert-butylcatechol

Product Code. 11382077
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11382077

Brand: Thermo Scientific Alfa Aesar L07860.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28469-92-3 | C8H6Cl2 | 173.04 g/mol

2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28469-92-3
Molecular Formula C8H6Cl2
Molecular Weight (g/mol) 173.04
MDL Number MFCD00000579
InChI Key YJCVRMIJBXTMNR-UHFFFAOYSA-N
Synonym 2,6-dichlorostyrene, 1,3-dichloro-2-vinylbenzene, benzene, 1,3-dichloro-2-ethenyl, 2,6-dichlorstyrol, acmc-20ao92, 1,3-dichloro-2-vinylbenzene #, 2,6-dichlorostyrene, stabilized 5g
PubChem CID 34254
IUPAC Name 1,3-dichloro-2-ethenylbenzene
SMILES ClC1=CC=CC(Cl)=C1C=C

Specifications

Density 1.266
Boiling Point 88°C to 90°C (8 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.574
Quantity 25 g
Beilstein 1931527
Solubility Information Immiscible with water.
Formula Weight 173.04
Percent Purity 96%
Chemical Name or Material 2,6-Dichlorostyrene, stabilized with 0.1% 4-tert-butylcatechol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.