missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-nitrotoluene, 98%, Thermo Scientific™

Product Code. 15480119
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15480119

Brand: Thermo Scientific Alfa Aesar H63431.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluoro-3-nitrotoluene used as a intermediate in organic synthesis, pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79562-49-5
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD11519030
InChI Key AVWNNVJXXMKAPB-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene
PubChem CID 12795486
IUPAC Name 1,3-difluoro-2-methyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1F)[N+](=O)[O-])F

Specifications

Quantity 250 mg
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 173.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,6-Difluoro-3-nitrotoluene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.