missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-hydroxybenzoic acid, 95%

Product Code. 15433407
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15433407

Brand: Thermo Scientific Alfa Aesar H26134.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 214917-68-7 | C7H4F2O3 | 174.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214917-68-7
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.10
MDL Number MFCD03094500
InChI Key NFIQGYBXSJQLSR-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol
PubChem CID 2778776
IUPAC Name 2,6-difluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1F

Specifications

Melting Point 170°C to 171°C (decomposition)
Quantity 250 mg
Formula Weight 174.1
Percent Purity 95%
Chemical Name or Material 2,6-Difluoro-4-hydroxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.