missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-iodophenol, 99%

Product Code. 11390026
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390026 1 g 1g
11380026 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390026 Supplier Thermo Scientific Alfa Aesar Supplier No. H32080.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 950858-06-7 | C6H3F2IO | 255.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 950858-06-7
Molecular Formula C6H3F2IO
Molecular Weight (g/mol) 255.99
MDL Number MFCD09907951
InChI Key LHUADYKKTXZZMQ-UHFFFAOYSA-N
PubChem CID 45790937
IUPAC Name 2,6-difluoro-4-iodophenol
SMILES C1=C(C=C(C(=C1F)O)F)I

Specifications

Melting Point 70°C to 73°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 255.99
Percent Purity 99%
Chemical Name or Material 2,6-Difluoro-4-iodophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.