missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoroaniline, TRC

Product Code. 30006211
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006211 5 g 5g
30007384 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006211 Supplier Toronto Research Chemicals Supplier No. TRCD4455305G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5509-65-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
Synonym 2,6-Difluorobenzenamine, 2,6-Difluorobenzeneamine, 2,6-Difluorophenylamine, NSC 127234;
SMILES Nc1c(F)cccc1F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H5F2N/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2
Formula Weight 129.039
Chemical Name or Material 2,6-Difluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.