missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF

Product Code. 15460876
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460876 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460876 Supplier Thermo Scientific Alfa Aesar Supplier No. H54189.AE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107549-26-8 | C7H5BrF2Mg | 231.32 g/mol

2,6-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store under inert gas. Store away from
TRUSTED_SUSTAINABILITY

Specifications

CAS 107549-26-8
Molecular Formula C7H5BrF2Mg
MDL Number MFCD11113508
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 2,6-difluorobenzylmagnesium bromide, 0.25m in 2-methf, 2,6-difluorobenzylmagnesium bromide, grignard reagent
Solubility Information Reacts with water.
InChI Key KEOOIBWMYNKEEF-UHFFFAOYSA-M
SMILES FC1=CC=CC(F)=C1C[Mg]Br
IUPAC Name bromo[(2,6-difluorophenyl)methyl]magnesium
Molecular Weight (g/mol) 231.32
PubChem CID 73995681
Formula Weight 231.32
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 2,6-Difluorobenzylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.