missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenol, 98+%, Thermo Scientific™

Product Code. 11478676
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11478676

Brand: Thermo Scientific Alfa Aesar A16034.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluorophenol undergoes oxidative polymerization in the presence of the Fe-N,N′-bis(salicylidene)ethylenediamine (salen) complex (catalyst) and hydrogen peroxide (oxidizing agent) to give poly(2,6-difluoro-1,4-phenylene oxide). It is also used as pharmaceutical intermediates.

Solubility
Slightly soluble in water. Soluble in ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28177-48-2
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00002158
InChI Key CKKOVFGIBXCEIJ-UHFFFAOYSA-N
Synonym phenol, 2,6-difluoro, 2.6-difluorophenol, 2,6-difluoro-phenol, 2,6-difluorophenol, 2,6-difluoro phenol, pubchem1495, ksc493c1n, ckkovfgibxceij-uhfffaoysa
PubChem CID 94392
IUPAC Name 2,6-difluorophenol
SMILES C1=CC(=C(C(=C1)F)O)F

Specifications

Melting Point 36°C to 40°C
Density 1.27
Boiling Point 59°C to 61°C (17 mmHg)
Flash Point 58°C (136°F)
Quantity 25 g
UN Number UN2921
Beilstein 2043613
Solubility Information Slightly soluble in water. Soluble in ethanol.
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 2,6-Difluorophenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.