missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenylacetone, 98%, Thermo Scientific™

Product Code. 10260303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10260303 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10260303 Supplier Thermo Scientific Acros Supplier No. 379150010
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 101712-20-3
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H8F2O
Linear Formula (F)2C6H3CH2COCH3
Quantity 1 g
Synonym 1-2,6-difluorophenyl propan-2-one, 2-propanone, 1-2,6-difluorophenyl, 2,6-difluorophenylacetone
InChI Key LAWHOFKPDMZDLJ-UHFFFAOYSA-N
SMILES CC(=O)CC1=C(C=CC=C1F)F
IUPAC Name 1-(2,6-difluorophenyl)propan-2-one
Molecular Weight (g/mol) 170.16
PubChem CID 2734927
Formula Weight 170.16
Percent Purity 98%
Chemical Name or Material 2,6-Difluorophenylacetone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.