missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylphenol, 97%

Product Code. 11397006
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397006 25 g 25g
11307016 100 g 100g
11317016 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397006 Supplier Thermo Scientific Alfa Aesar Supplier No. L06841.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An anesthetic that exhibits direct activation of the GABA-A receptor | CAS: 2078-54-8 | C12H18O | 178.275 g/mol

2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

Solubility
Very slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2078-54-8
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00008885
InChI Key OLBCVFGFOZPWHH-UHFFFAOYSA-N
Synonym propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum
PubChem CID 4943
ChEBI CHEBI:44915
IUPAC Name 2,6-di(propan-2-yl)phenol
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)O

Specifications

Melting Point 17°C to 18°C
Density 0.962g/mL
Boiling Point 254°C to 256°C
Refractive Index 1.5140
Quantity 25 g
Beilstein 1866484
Merck Index 14,7834
Formula Weight 178.28
Percent Purity 97%
Chemical Name or Material 2,6-Diisopropylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.