missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylphenol, 97%, Thermo Scientific™

An anesthetic that exhibits direct activation of the GABA-A receptor

34.99 € - 312.99 €

Chemical Identifiers

CAS 2078-54-8
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00008885
InChI Key OLBCVFGFOZPWHH-UHFFFAOYSA-N
Synonym propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum
PubChem CID 4943
ChEBI CHEBI:44915
IUPAC Name 2,6-di(propan-2-yl)phenol
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397006
SDS View Documents
Thermo Scientific Alfa Aesar
L06841.14
25g
ONLINE EXCLUSIVE PRICE
36.80 €
34.99 € / 25g
Save 1.81 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11307016
SDS View Documents
Thermo Scientific Alfa Aesar
L06841.22
100g
ONLINE EXCLUSIVE PRICE
95.50 €
93.59 € / 100g
Save 1.91 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11317016
SDS View Documents
Thermo Scientific Alfa Aesar
L06841.36
500g
ONLINE EXCLUSIVE PRICE
319.00 €
312.99 € / 500g
Save 6.01 €
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

Solubility
Very slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2078-54-8
178.275
OLBCVFGFOZPWHH-UHFFFAOYSA-N
4943
2,6-di(propan-2-yl)phenol
C12H18O
MFCD00008885
propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum
CHEBI:44915
CC(C)C1=C(C(=CC=C1)C(C)C)O

Specifications

2078-54-8
254°C to 256°C
1.5140
1866484
propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum
CC(C)C1=C(C(=CC=C1)C(C)C)O
178.275
CHEBI:44915
97%
0.962g/mL
C12H18O
MFCD00008885
14,7834
OLBCVFGFOZPWHH-UHFFFAOYSA-N
2,6-di(propan-2-yl)phenol
4943
178.28
2,6-Diisopropylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'rtecsNumber' : SL0810000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.