missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxy-4-methylphenol, 97%

Product Code. 11434817
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11434817

Brand: Thermo Scientific Alfa Aesar B21801.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6638-05-7 | C9H12O3 | 168.19 g/mol

Used as a flavor and fragrance agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavor and fragrance agents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6638-05-7
MDL Number MFCD00017289

Specifications

Melting Point 36°C to 42°C
Boiling Point 144°C to 146°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Phenol-like
Molecular Formula C16H13NO3
Quantity 25 g
Synonym 5-benzyloxy-1h-indole-2-carboxylic acid, 5-benzyloxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-phenylmethoxy, chembl24850, 5-phenylmethoxy-1h-indole-2-carboxylic acid, 5-phenylmethoxy indole-2-carboxylic acid, 5-benzyloxy-1h-indole-2-carboxylicacid, maybridge1_005333, acmc-209x2s
Solubility Information Slightly soluble in water.
InChI Key MVCLSAMNMAWXFQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC(=C3)C(=O)O
IUPAC Name 5-phenylmethoxy-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 267.28
PubChem CID 81146
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 2,6-Dimethoxy-4-methylphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.