missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyaniline, 96%

Product Code. 13428628 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13428628 5 g Glass bottle 5g
13418628 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13428628 Supplier Thermo Scientific Acros Supplier No. 448790050
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2734-70-5 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2734-70-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
InChI Key HQBJSEKQNRSDAZ-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dimethoxy, 2,6-dimethoxyphenylamine, 2,6-dimethoxybenzenamine, 2,6-dimethoxy-phenylamine, buttpark 33\04-69, 2 6-dimethoxyaniline, pubchem4419, 2,6-dimethoxy aniline, acmc-1cnwd, ksc497k1h
PubChem CID 95940
IUPAC Name 2,6-dimethoxyaniline
SMILES COC1=C(C(=CC=C1)OC)N

Specifications

Melting Point 76°C to 78°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3NH2
Quantity 5 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 153.18
Percent Purity 96%
Chemical Name or Material 2, 6-Dimethoxyaniline
compliance-icons
Product Identifier
  • 2,6-Dimethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.