missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyaniline, 97%

Product Code. p-7029650
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11456250

Brand: Thermo Scientific Alfa Aesar H27139.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2734-70-5 | C8H11NO2 | 153.181 g/mol

2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2734-70-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00053934
InChI Key HQBJSEKQNRSDAZ-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dimethoxy, 2,6-dimethoxyphenylamine, 2,6-dimethoxybenzenamine, 2,6-dimethoxy-phenylamine, buttpark 33\04-69, 2 6-dimethoxyaniline, pubchem4419, 2,6-dimethoxy aniline, acmc-1cnwd, ksc497k1h
PubChem CID 95940
IUPAC Name 2,6-dimethoxyaniline
SMILES COC1=C(C(=CC=C1)OC)N

Specifications

Melting Point 76°C to 78°C
Odor Characteristic
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 153.18
Percent Purity 97%
Chemical Name or Material 2,6-Dimethoxyaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.