missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyphenol, 99%

Product Code. 11411307
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11411307

Brand: Thermo Scientific Alfa Aesar B24253.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-10-1 | C8H10O3 | 154.17 g/mol

It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-10-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064434
InChI Key KLIDCXVFHGNTTM-UHFFFAOYSA-N
Synonym syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol
PubChem CID 7041
ChEBI CHEBI:955
IUPAC Name 2,6-dimethoxyphenol
SMILES COC1=CC=CC(OC)=C1O

Specifications

Melting Point 50°C to 57°C
Boiling Point 261°C
Flash Point 140°C (284°F)
Quantity 25 g
Beilstein 1526871
Solubility Information Soluble in water.
Formula Weight 154.16
Percent Purity 99%
Chemical Name or Material 2,6-Dimethoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.