missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline, 99%

Product Code. 10172570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10172570 5 g Glass bottle 5g
10162620 100 g Glass bottle 100g
10753651 500 g Glass bottle 500g
10700965 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10172570 Supplier Thermo Scientific Acros Supplier No. 115940050
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-62-7 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-62-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007747
InChI Key UFFBMTHBGFGIHF-UHFFFAOYSA-N
Synonym 2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
PubChem CID 6896
ChEBI CHEBI:28738
IUPAC Name 2,6-dimethylaniline
SMILES CC1=CC=CC(C)=C1N

Specifications

Melting Point 10°C to 12°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9800g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5591 to 1.5611
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.98
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1107
Merck Index 15, 10282
Boiling Point 216°C
Solubility Information Solubility in water: 7.5g/L (20 c). Other solubilities: very soluble in ether and ethanol,soluble in dimethyl sulfoxide,carbon tetrachlorid
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2, 6-Dimethylaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Xylidene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.