missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline, TRC

Product Code. 17944305
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17944305 100 g 100g
17904205 10 g 10g
17983315 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944305 Supplier Toronto Research Chemicals Supplier No. TRCD462195100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-62-7
Molecular Formula C8 H11 N
Molecular Weight (g/mol) 121.18
Synonym 2,6-Xylidine (8CI), 2,6-Dimethylbenzenamine, 1-Amino-2,6-dimethylbenzene, 2,6-Dimethylaminobenzene, 2,6-Dimethylaniline, 2,6-Dimethylphenylamine, 2,6-Xylylamine, 2-Amino-1,3-dimethylbenzene, 2-Amino-1,3-xylene, 2-Amino-m-xylene, NSC 7098, o-Xylidine
IUPAC Name 2,6-dimethylaniline
SMILES Cc1cccc(C)c1N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3
Formula Weight 121.0891
Percent Purity >95%
Chemical Name or Material 2,6-Dimethylaniline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.