missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylbenzaldehyde, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
An aldehyde containing building block used for proteomics research
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An aldehyde containing building block used for proteomics research
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
An aldehyde containing building block used for proteomics research
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 27°C to 30°C (literature) |
| Color | White to Yellow |
| Boiling Point | 230°C (literature) |
| Flash Point | 96°C (205°F) |
| Refractive Index | 1.548 |
| Quantity | 25 g |
| Beilstein | 907019 |
| Sensitivity | Air Sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 134.18 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction