missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenol, 99%

Product Code. 10540404 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
2.5kg
250g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10540404 5 kg Plastic Bottle 5kg
10447300 5 g Glass bottle 5g
10447490 250 g Glass bottle 250g
10050730 1 kg Glass bottle 1kg
10052823 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10540404 Supplier Thermo Scientific Acros Supplier No. 116570051
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 576-26-1 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-26-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002240
InChI Key NXXYKOUNUYWIHA-UHFFFAOYSA-N
Synonym 2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
PubChem CID 11335
IUPAC Name 2,6-dimethylphenol
SMILES CC1=CC=CC(C)=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 45.0°C to 48.0°C
Color Brown-Yellow to Colorless
Density 1.1500g/mL
Boiling Point 203.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Bottle
Linear Formula (CH3)2C6H3OH
Quantity 5 kg
Beilstein 06, 485
Merck Index 15, 10279
Specific Gravity 1.15
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
Formula Weight 122.17
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2, 6-Dimethylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.