missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitroaniline, 95%

Product Code. 13515000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13515000 10 g Glass bottle 10g
13584990 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13515000 Supplier Thermo Scientific Acros Supplier No. 170170100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 606-22-4 | C6H5N3O4 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-22-4
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.12
InChI Key QFUSCYRJMXLNRB-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
PubChem CID 69070
IUPAC Name 2,6-dinitroaniline
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 137.0°C to 142.0°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula (O2N)2C6H3NH2
Quantity 10 g
Beilstein 12,758
Merck Index 15,3297
Solubility Information Solubility in water: insoluble. Other solubilities: 0.4g/100mL ethanol 95%,soluble in ether and hot benzene,practically insoluble in petroleum ether
Formula Weight 183.12
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 6-Dinitroaniline, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dinitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.