missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diaminofluorene, 97%

Product Code. 15461628
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461628 1 g 1g
15481628 250 mg 250mg
15471628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461628 Supplier Thermo Scientific Alfa Aesar Supplier No. H55402.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 525-64-4 | C13H13ClN2 | 232.71 g/mol

It is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.

Solubility
Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether.

Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-64-4
Molecular Formula C13H13ClN2
Molecular Weight (g/mol) 232.71
MDL Number MFCD00001128
InChI Key COOWDVCIUYIBFE-UHFFFAOYSA-N
Synonym 2,7-diaminofluorene, 2,7-fluorenediamine, 2,7-diamino-fluorene, 2,7-fluoroenediamine, fluorene-2,7-diamine, fluorene-2,7-diyldiamine, ccris 920, 2,7-diamino-fluoren, 2,7-diaminoflyorene, 2,7-diaminofluoren
PubChem CID 10679
IUPAC Name 9H-fluorene-2,7-diamine
SMILES [Cl-].NC1=CC=C2C(CC3=CC([NH3+])=CC=C23)=C1

Specifications

Melting Point 160°C to 162°C
Quantity 1 g
Beilstein 2099859
Merck Index 14,4156
Solubility Information Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether.
Formula Weight 196.25
Percent Purity ≥97%
Chemical Name or Material 2,7-Diaminofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.