missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diisopropyl-9,9-dimethyl-9,10-dihydroacridine, TRC

Product Code. 30384766
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384766 10 mg 10mg
30376035 50 mg 50mg
30372162 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384766 Supplier Toronto Research Chemicals Supplier No. TRCD45576010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,7-Diisopropyl-9,9-dimethyl-9,10-dihydroacridine
Analyte or Component Names 2,7-Diisopropyl-9,9-dimethyl-9,10-dihydroacridine
Shipping Condition Room Temperature
Molecular Formula C21 H29 N
Synonym Diisopropyldimethylacridine
InChI Formula InChI=1S/C21H29N/c1-13(2)15-7-9-19-17(11-15)21(5,6)18-12-16(14(3)4)8-10-20(18)22-19/h7-14,17,19,22H,1-6H3
Solution Type Neat
SMILES CC(C)C1=CC2C(Nc3ccc(cc3C2(C)C)C(C)C)C=C1
IUPAC Name 9,9-dimethyl-2,7-di(propan-2-yl)-9a,10-dihydro-4aH-acridine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 295.462
Quantity 10 mg
Formula Weight 295.23
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.