missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dinitro-9-fluorenone, 97%

Product Code. 15481338
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481338 25 g 25g
15471338 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481338 Supplier Thermo Scientific Alfa Aesar Supplier No. H55176.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31551-45-8 | C13H6N2O5 | 270.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31551-45-8
Molecular Formula C13H6N2O5
Molecular Weight (g/mol) 270.20
MDL Number MFCD00001153
InChI Key HDVGAFBXTXDYIB-UHFFFAOYSA-N
Synonym 2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
PubChem CID 35842
IUPAC Name 2,7-dinitrofluoren-9-one
SMILES [O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O

Specifications

Melting Point 292°C to 295°C
Color Yellow to Orange
Quantity 25 g
Formula Weight 270.2
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2,7-Dinitro-9-fluorenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.