missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 10590712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10590712 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10590712 Supplier Thermo Scientific Acros Supplier No. 354020050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38985-79-4
Molecular Formula C9H7BrNO3
Molecular Weight (g/mol) 257.06
MDL Number MFCD00040904
InChI Key QVABAFHRLMDDLM-UHFFFAOYSA-M
PubChem CID 520935
SMILES CC(=O)NC1=CC=C(Br)C=C1C([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 218°C to 223°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C(o)NHC6H3(Br)COOH
Quantity 5 g
Synonym 5-bromo-2-acetamidobenzoic acid, 2-acetamino-5-bromobenzoic acid, n-acetyl-5-bromo-anthranilic acid, 2-acetylamino-5-bromobenzoic acid, n-acetyl-5-bromoanthranilic acid, 2-acetamido-5-bromobenzoic acid, benzoic acid,2-acetylamino-5-bromo, benzoic acid, 2-acetylamino-5-bromo
IUPAC Name 2-acetamido-5-bromobenzoic acid
Formula Weight 258.07
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Acetamido-5-bromobenzoic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.