missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Acetamido-5-bromobenzoic acid, 98%

Product Code. 11341527
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341527 5 g 5g
11351527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341527 Supplier Alfa Aesar™ Supplier No. L02139.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 38985-79-4
Molecular Formula C9H7BrNO3
Molecular Weight (g/mol) 257.06
MDL Number MFCD00040904
InChI Key QVABAFHRLMDDLM-UHFFFAOYSA-M
PubChem CID 520935
IUPAC Name 2-acetamido-5-bromobenzoic acid
SMILES CC(=O)NC1=CC=C(Br)C=C1C([O-])=O

Specifications

Melting Point 222°C to 224°C
Quantity 5 g
Beilstein 882463
Synonym 5-bromo-2-acetamidobenzoic acid, 2-acetamino-5-bromobenzoic acid, n-acetyl-5-bromo-anthranilic acid, 2-acetylamino-5-bromobenzoic acid, n-acetyl-5-bromoanthranilic acid, 2-acetamido-5-bromobenzoic acid, benzoic acid,2-acetylamino-5-bromo, benzoic acid, 2-acetylamino-5-bromo
Formula Weight 258.07
Percent Purity 98%
Chemical Name or Material 2-Acetamido-5-bromobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.