missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidophenol, 97%

Product Code. 11484797
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484797 100 g 100g
11474797 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484797 Supplier Thermo Scientific Alfa Aesar Supplier No. B21264.22
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 614-80-2 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-80-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002181
InChI Key ADVGKWPZRIDURE-UHFFFAOYSA-N
Synonym 2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
PubChem CID 11972
IUPAC Name N-(2-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1O

Specifications

Melting Point 205°C to 210°C
Quantity 100 g
Beilstein 607592
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 2-Acetamidophenol
compliance-icons
Product Identifier
  • 2-Acetamidophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: N-(2-hydroxyphenyl)-acetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.