missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyrazine, 99%

Product Code. 11498446
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498446 5 g 5g
11488446 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498446 Supplier Thermo Scientific Alfa Aesar Supplier No. A15917.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22047-25-2 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22047-25-2
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006134
InChI Key DBZAKQWXICEWNW-UHFFFAOYSA-N
Synonym acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone
PubChem CID 30914
IUPAC Name 1-pyrazin-2-ylethanone
SMILES CC(=O)C1=NC=CN=C1

Specifications

Melting Point 75°C to 78°C
Boiling Point 78°C to 79°C (8 mmHg)
Quantity 5 g
Beilstein 109630
Formula Weight 122.13
Percent Purity 99%
Chemical Name or Material 2-Acetylpyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.