accessibility menu, dialog, popup

UserName

H3CONN

CAS RN 22047-25-2

IUPAC Name: 1-(pyrazin-2-yl)ethan-1-one
Synonyms: acetylpyrazine
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: DBZAKQWXICEWNW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CN=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)