missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-4,6-dibenzoylresorcinol, 98%

Product Code. 11379495
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379495 5 g 5g
11389495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379495 Supplier Thermo Scientific Alfa Aesar Supplier No. L18722.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 102593-74-8 | C23H18O4 | 358.393 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102593-74-8
Molecular Formula C23H18O4
Molecular Weight (g/mol) 358.393
MDL Number MFCD02094038
InChI Key FSYGSBMXRNPJAD-UHFFFAOYSA-N
Synonym 2-allyl-4,6-dibenzoylresorcinol, 5-allyl-4,6-dihydroxy-1,3-phenylene bis phenylmethanone, methanone, 4,6-dihydroxy-5-2-propenyl-1,3-phenylene bis phenyl, 4,6-dibenzoyl-2-prop-2-en-1-yl benzene-1,3-diol, 2allyl-4,6-dibenzoylresorcinol, methanone, 1,1'-4,6-dihydroxy-5-2-propen-1-yl-1,3-phenylene bis 1-phenyl, pubchem21315, acmc-20ao5t
PubChem CID 7010346
IUPAC Name (5-benzoyl-2,4-dihydroxy-3-prop-2-enylphenyl)-phenylmethanone
SMILES C=CCC1=C(C(=CC(=C1O)C(=O)C2=CC=CC=C2)C(=O)C3=CC=CC=C3)O

Specifications

Melting Point 158°C to 163°C
Quantity 5 g
Beilstein 2168154
Formula Weight 358.4
Percent Purity 98%
Chemical Name or Material 2-Allyl-4,6-dibenzoylresorcinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.