missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-D-glycine, 95%

Product Code. 15408637
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15408637

Brand: Thermo Scientific Alfa Aesar H52112.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54594-06-8 | C5H9NO2 | 115.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54594-06-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00063104
InChI Key WNNNWFKQCKFSDK-SCSAIBSYSA-N
Synonym d-allylglycine, r-2-aminopent-4-enoic acid, r-2-amino-4-pentenoic acid, 2r-2-aminopent-4-enoic acid, h-d-gly allyl-oh, r-2-amino-pent-4-enoic acid, 4-pentenoic acid, 2-amino-, 2r, 2r-2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, r-allylglycine
PubChem CID 6992334
IUPAC Name (2R)-2-aminopent-4-enoic acid
SMILES C=CCC(C(=O)O)N

Specifications

Quantity 1 g
Formula Weight 115.13
Percent Purity 95%
Chemical Name or Material 2-Allyl-D-glycine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.