missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-N-Fmoc-L-glycine, 95%

Product Code. 15478727
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478727 5 g 5g
15488727 250 mg 250mg
15468727 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15478727 Supplier Thermo Scientific Alfa Aesar Supplier No. H52177.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 146549-21-5 | C20H19NO4 | 337.375 g/mol

2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146549-21-5
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
MDL Number MFCD01311749
InChI Key YVBLQCANYSFEBN-SFHVURJKSA-N
Synonym fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine
PubChem CID 2734457
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
SMILES C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 5 g
Beilstein 5884047
Solubility Information Slightly soluble in water.
Formula Weight 337.37
Percent Purity 95%
Chemical Name or Material 2-Allyl-N-Fmoc-L-glycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.