missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-naphthalenesulfonic Acid, TRC

Product Code. 30132538
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132538 100 g 100g
30134956 250 g 250g
17938334 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132538 Supplier Toronto Research Chemicals Supplier No. TRCA618330100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-16-3
Formula Weight 223.0303
InChI Formula InChI=1S/C10H9NO3S/c11-9-6-5-7-3-1-2-4-8(7)10(9)15(12,13)14/h1-6H,11H2,(H,12,13,14)
IUPAC Name 2-aminonaphthalene-1-sulfonic acid
Molecular Formula C10 H9 N O3 S
Molecular Weight (g/mol) 223.25
Percent Purity >95%
SMILES Nc1ccc2ccccc2c1S(=O)(=O)O
Synonym Tobias Acid, 2-Naphthylamine-1-sulfonic Acid, 1-Sulfo-2-naphthylamine
Chemical Name or Material 2-Amino-1-naphthalenesulfonic Acid
Grade HPLC
Show More Show Less

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.