missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1,3-propanediol, 99+%

Product Code. 15475409
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475409 25 g 25g
15485409 100 g 100g
15495409 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475409 Supplier Thermo Scientific Alfa Aesar Supplier No. J63144.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A useful biological buffer | CAS: 115-69-5 | C4H11NO2 | 105.137 g/mol

A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

Solubility
Soluble in water and other organic solvents.

Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, aluminum, copper, brass and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-69-5
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00004678
InChI Key UXFQFBNBSPQBJW-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
PubChem CID 1531
ChEBI CHEBI:991
IUPAC Name 2-amino-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)N

Specifications

Melting Point 100°C to 110°C
Color White
Boiling Point 151°C (10 mmHg)
Linear Formula (HOCH2)2C(NH2)CH3
Quantity 25 g
Beilstein 635708
Merck Index 14,448
Solubility Information Soluble in water and other organic solvents.
Formula Weight 105.14
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 2-Amino-2-methyl-1,3-propanediol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-2-methyl-1,3-propanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: 2-Amino-2-methyl-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.