missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methylpropane-1-sulfonic acid, TRC

Product Code. 30006688
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006688 10 mg 10mg
30006722 100 mg 100mg
30008618 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006688 Supplier Toronto Research Chemicals Supplier No. TRCA89869510MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86311-35-5
Molecular Formula C4 H11 N O3 S
Molecular Weight (g/mol) 153.2
IUPAC Name 2-amino-2-methylpropane-1-sulfonic acid
SMILES CC(C)(N)CS(=O)(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H11NO3S/c1-4(2,5)3-9(6,7)8/h3,5H2,1-2H3,(H,6,7,8)
Formula Weight 153.046
Chemical Name or Material 2-Amino-2-methylpropane-1-sulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.