missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,4-dimethylbenzoic acid, 98+%

Product Code. 11496986
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496986 5 g 5g
11406996 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496986 Supplier Thermo Scientific Alfa Aesar Supplier No. B25313.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50419-58-4 | C9H11NO2 | 165.19 g/mol

2-Amino-3,4-dimethylbenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,4-dimethylbenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50419-58-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00130041
InChI Key MUOBMUYSNYMSDM-UHFFFAOYSA-N
Synonym 2-amino-3,4-dimethyl-benzoic acid, 3,4-dimethylanthranilic acid, benzoic acid, 2-amino-3,4-dimethyl, pubchem4683, acmc-1aomr, ksc272a3h, 2-amino-3,4-dimethyl benzoic acid
PubChem CID 282450
IUPAC Name 2-amino-3,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(N)=C1C

Specifications

Melting Point ∼176°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 165.19
Percent Purity ≥98%
Chemical Name or Material 2-Amino-3,4-dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.