missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11397086
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397086 5 g 5g
11387086 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397086 Supplier Thermo Scientific Alfa Aesar Supplier No. L07003.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14438-32-5 | Molecular Formula: C9H11NO2 | Molecular Weight: 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14438-32-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017099
InChI Key GIMYRAQQQBFFFJ-UHFFFAOYSA-N
Synonym 3,5-dimethylanthranilic acid, 2-amino-3,5-dimethyl-benzoic acid, 2-amino-3,5-dimethyl benzoic acid, benzoic acid, 2-amino-3,5-dimethyl, pubchem4685, 3,5-dimethylanthranilic, acmc-1bqlm, intermediates-zcf02106, bidd:gt0362, 2-amino-3,5-dimethylbenzoicacid
PubChem CID 259834
IUPAC Name 2-amino-3,5-dimethylbenzoic acid
SMILES CC1=CC(=C(C(=C1)C(=O)O)N)C

Specifications

Melting Point 189°C to 194°C
Quantity 5 g
Beilstein 1101785
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.