missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. 11323406
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323406 5 g 5g
11313406 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323406 Supplier Thermo Scientific Alfa Aesar Supplier No. L12850.06
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3032-32-4 | Molecular Formula: C7H5Cl2NO2 | Molecular Weight: 206.02 g/mol

2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, bases and sodium hypochlorite.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3032-32-4
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00052390
InChI Key MMIREQFMYZQWLU-UHFFFAOYSA-N
PubChem CID 604542
IUPAC Name 2-amino-3,6-dichlorobenzoic acid
SMILES NC1=C(Cl)C=CC(Cl)=C1C(O)=O

Specifications

Melting Point 150°C to 154°C
Quantity 5 g
Beilstein 3266879
Synonym benzoic acid, 2-amino-3,6-dichloro, 3,6-dichloroanthranilic acid, 2-amino-3,6-dichlorobenzoicacid, acmc-1afqi, 6-amino-2,5-dichlorobenzoic acid, benzoic acid,2-amino-3,6-dichloro, 2-azanyl-3,6-bis chloranyl benzoic acid
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 95%
Chemical Name or Material 2-Amino-3,6-dichlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.