missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. p-7046025
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11313406

Brand: Thermo Scientific Alfa Aesar L12850.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, bases and sodium hypochlorite.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3032-32-4
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00052390
InChI Key MMIREQFMYZQWLU-UHFFFAOYSA-N
PubChem CID 604542
IUPAC Name 2-amino-3,6-dichlorobenzoic acid
SMILES NC1=C(Cl)C=CC(Cl)=C1C(O)=O

Specifications

Melting Point 150°C to 154°C
Quantity 1 g
Beilstein 3266879
Synonym benzoic acid, 2-amino-3,6-dichloro, 3,6-dichloroanthranilic acid, 2-amino-3,6-dichlorobenzoicacid, acmc-1afqi, 6-amino-2,5-dichlorobenzoic acid, benzoic acid,2-amino-3,6-dichloro, 2-azanyl-3,6-bis chloranyl benzoic acid
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 95%
Chemical Name or Material 2-Amino-3,6-dichlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.