missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,8-dimethylimidazo[4,5-f]quinoxaline, TRC

Product Code. 30005956
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005956 5 mg 5mg
30004635 50 mg 50mg
30006570 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005956 Supplier Toronto Research Chemicals Supplier No. TRCA6066005MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77500-04-0
Molecular Formula C11 H11 N5
Molecular Weight (g/mol) 213.24
Synonym 3H-Imidazo[4,5-f]quinoxalin-2-amine, 3,8-dimethyl-, 3,8-Dimethyl-3H-imidazo[4,5-f]quinoxalin-2-amine, 2-Amino-3,8-dimethylimidazo[4,5-f]quinoxaline, 2-Amino-3,8-dimethylmidazo[4,5-f]quinoxaline, MeIQx
IUPAC Name 3,8-dimethylimidazo[4,5-f]quinoxalin-2-amine
SMILES Cc1cnc2ccc3c(nc(N)n3C)c2n1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H11N5/c1-6-5-13-7-3-4-8-10(9(7)14-6)15-11(12)16(8)2/h3-5H,1-2H3,(H2,12,15)
Formula Weight 213.1014
Percent Purity >95%
Chemical Name or Material 2-Amino-3,8-dimethylimidazo[4,5-f]quinoxaline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.