missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-methylbenzoic acid, 98+%

Product Code. 11452797
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452797 5 g 5g
11339699 1 g 1g
11462797 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452797 Supplier Thermo Scientific Alfa Aesar Supplier No. A15443.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13091-43-5 | C8H8BrNO2 | 230.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13091-43-5
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00051705
InChI Key LCMZECCEEOQWLQ-UHFFFAOYSA-N
PubChem CID 2774400
IUPAC Name 2-amino-3-bromo-5-methylbenzoic acid
SMILES CC1=CC(Br)=C(N)C(=C1)C(O)=O

Specifications

Melting Point 202°C to 206°C
Quantity 5 g
Beilstein 3540992
Synonym 3-bromo-5-methylanthranilic acid, 2-amino-3-bromo-5-methyl-benzoic acid, 6-amino-5-bromo-m-toluic acid, benzoic acid, 2-amino-3-bromo-5-methyl, acmc-20ao5e, 4-amino-3-bromo-5-carboxytoluene, 2-bromo-6-carboxy-4-methylaniline, benzoic acid,2-amino-3-bromo-5-methyl, 2-azanyl-3-bromanyl-5-methyl-benzoic acid, 2-amino-3-bromo-5-methylbenzoic acid
Formula Weight 230.07
Percent Purity ≥98%
Chemical Name or Material 2-Amino-3-bromo-5-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.