missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-nitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 10480774
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10480774 50 g Amber Glass Bottle 50g
10576134 100 g Amber Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10480774 Supplier Thermo Scientific Maybridge Supplier No. SB01878EE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17601-94-4
Molecular Formula C7H4BrN3O2
Molecular Weight (g/mol) 242.032
InChI Key MUHLVSZIVTURCZ-UHFFFAOYSA-N
Synonym benzonitrile, 2-amino-3-bromo-5-nitro, 6-bromo-2-cyano-4-nitroaniline, 2-bromo-6-cyano-4-nitroaniline, 2-cyano-4-nitro-6-bromoaniline, 2-amino-3-bromo-5-nitrobenzenecarbonitrile, pubchem2331, 2-amino-3-bromo-1-cyano-5-nitrobenzene, acmc-1bus8, cambridge id 5228140, ksc496g6h
PubChem CID 87173
IUPAC Name 2-amino-3-bromo-5-nitrobenzonitrile
SMILES C1=C(C=C(C(=C1Br)N)C#N)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 242.03
Percent Purity 97%
Chemical Name or Material 2-Amino-3-bromo-5-nitrobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.