missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chlorobenzoic acid, 97%, Thermo Scientific™

Product Code. 10447012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10447012 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10447012 Supplier Thermo Scientific Acros Supplier No. 335970010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6388-47-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 190°C to 195°C
Color Gray or Tan-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H6ClNO2
Linear Formula C6H3ClNH2COOH
MDL Number MFCD00134229
Quantity 1 g
Synonym 3-chloroanthranilic acid, benzoic acid, 2-amino-3-chloro, 2-amino-3-chloro-benzoic acid, 3-chloro-2-aminobenzoic acid, anthranilic acid, 3-chloro, 2-amino-3-chlorobenzoicacid, pubchem4639, 3-chloroanthranilicacid, acmc-209nj4, amino-3-chlorobenzoic acid
InChI Key LWUAMROXVQLJKA-UHFFFAOYSA-N
SMILES NC1=C(Cl)C=CC=C1C(O)=O
IUPAC Name 2-amino-3-chlorobenzoic acid
Molecular Weight (g/mol) 171.58
PubChem CID 80807
Formula Weight 171.58
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chlorobenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.